Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTGTAGGATTTGTTGAACTTGT[C/T]GAAGGTGAAGTCCACAGTGTGCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147561 | ||||||||||||||||||||
Literature Links: |
ITGB5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGB5 - integrin subunit beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002213.4 | 2438 | Missense Mutation | AAC,GAC | N,D 788 | NP_002204.2 | |
XM_005247436.2 | 2438 | Missense Mutation | AAC,GAC | N,D 758 | XP_005247493.1 | |
XM_006713630.2 | 2438 | Missense Mutation | AAC,GAC | N,D 743 | XP_006713693.1 | |
XM_017006351.1 | 2438 | Missense Mutation | AAC,GAC | N,D 680 | XP_016861840.1 | |
XM_017006352.1 | 2438 | Missense Mutation | AAC,GAC | N,D 680 | XP_016861841.1 | |
XM_017006353.1 | 2438 | Missense Mutation | AAC,GAC | N,D 680 | XP_016861842.1 | |
XM_017006354.1 | 2438 | Missense Mutation | AAC,GAC | N,D 672 | XP_016861843.1 | |
XM_017006355.1 | 2438 | Missense Mutation | AAC,GAC | N,D 650 | XP_016861844.1 |