Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACAAGTGTTGAGCACCATAATA[A/C]GTAGGTGCTCAATAAATACATGAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606739 MIM: 605931 | |||||||||||||||||||||||
Literature Links: |
OSBPL11 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
OSBPL11 - oxysterol binding protein like 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022776.4 | 4237 | UTR 3 | NP_073613.2 |
SNX4 - sorting nexin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |