Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTGCAGATGGAATCAAGGCCAG[A/G]AACAGAAATCAGAACTACCTGGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF639 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF639 - zinc finger protein 639 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303425.1 | 979 | Silent Mutation | AGA,AGG | R,R 78 | NP_001290354.1 | |
NM_001303426.1 | 979 | Silent Mutation | AGA,AGG | R,R 78 | NP_001290355.1 | |
NM_016331.2 | 979 | Silent Mutation | AGA,AGG | R,R 78 | NP_057415.1 | |
XM_017006550.1 | 979 | Silent Mutation | AGA,AGG | R,R 78 | XP_016862039.1 | |
XM_017006551.1 | 979 | Silent Mutation | AGA,AGG | R,R 78 | XP_016862040.1 | |
XM_017006552.1 | 979 | Silent Mutation | AGA,AGG | R,R 78 | XP_016862041.1 | |
XM_017006553.1 | 979 | Silent Mutation | AGA,AGG | R,R 78 | XP_016862042.1 |