Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTACAATTACCTAAATGTGCCTT[C/T]TTGGAAAGAAATTCCAGGGTCAAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
VPS8 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
VPS8 - VPS8, CORVET complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009921.2 | Intron | NP_001009921.1 | ||||
NM_015303.3 | Intron | NP_056118.2 | ||||
XM_005247251.3 | Intron | XP_005247308.1 | ||||
XM_005247253.4 | Intron | XP_005247310.1 | ||||
XM_006713556.3 | Intron | XP_006713619.1 | ||||
XM_011512599.2 | Intron | XP_011510901.1 | ||||
XM_011512600.2 | Intron | XP_011510902.1 | ||||
XM_011512601.2 | Intron | XP_011510903.1 | ||||
XM_017006040.1 | Intron | XP_016861529.1 | ||||
XM_017006041.1 | Intron | XP_016861530.1 |