Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCTCAGCCTCTCCACCTTCCTA[C/T]CATGGCTTGAAGCAAGGTTTTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVC2 - EvC ciliary complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166136.1 | Intron | NP_001159608.1 | ||||
NM_147127.4 | Intron | NP_667338.3 | ||||
XM_017007735.1 | Intron | XP_016863224.1 | ||||
XM_017007736.1 | Intron | XP_016863225.1 | ||||
XM_017007737.1 | Intron | XP_016863226.1 | ||||
XM_017007738.1 | Intron | XP_016863227.1 | ||||
XM_017007739.1 | Intron | XP_016863228.1 |