Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGCTGTAGAATTTATATGCCC[A/G]GACATTTTGAGGACCAACAGTATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0232 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0232 - KIAA0232 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100590.1 | Intron | NP_001094060.1 | ||||
NM_014743.2 | Intron | NP_055558.2 | ||||
XM_005248041.4 | Intron | XP_005248098.1 | ||||
XM_011513598.2 | Intron | XP_011511900.1 | ||||
XM_017008860.1 | Intron | XP_016864349.1 | ||||
XM_017008861.1 | Intron | XP_016864350.1 | ||||
XM_017008862.1 | Intron | XP_016864351.1 | ||||
XM_017008863.1 | Intron | XP_016864352.1 |