Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGAGACAACAGCTGGGTGACAA[C/T]TCTAGCCCACCGTTCTCTGCTATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613379 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMBL - carboxymethylenebutenolidase homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138809.3 | 2411 | UTR 3 | NP_620164.1 | |||
XM_017009034.1 | 2411 | Intron | XP_016864523.1 | |||
XM_017009035.1 | 2411 | Intron | XP_016864524.1 |