Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGATGGAAAAACAGGATCTATTC[C/T]ACATAGAATATGCTTCTTGAATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DROSHA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DROSHA - drosha ribonuclease III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100412.1 | 4738 | UTR 3 | NP_001093882.1 | |||
NM_013235.4 | 4738 | UTR 3 | NP_037367.3 | |||
XM_005248291.3 | 4738 | Intron | XP_005248348.1 | |||
XM_005248292.3 | 4738 | Intron | XP_005248349.1 | |||
XM_005248293.3 | 4738 | Intron | XP_005248350.1 | |||
XM_005248294.3 | 4738 | Intron | XP_005248351.1 | |||
XM_011514033.2 | 4738 | Intron | XP_011512335.1 | |||
XM_017009399.1 | 4738 | Intron | XP_016864888.1 | |||
XM_017009400.1 | 4738 | Intron | XP_016864889.1 | |||
XM_017009401.1 | 4738 | Intron | XP_016864890.1 | |||
XM_017009402.1 | 4738 | Intron | XP_016864891.1 |