Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAGTACTGTTTTCTTTGTTTTC[C/T]GTAGGTTCTGATATGTTTTGTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR70 - WD repeat domain 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018034.2 | Intron | NP_060504.1 | ||||
XM_005248318.3 | Intron | XP_005248375.1 | ||||
XM_011514061.2 | Intron | XP_011512363.1 | ||||
XM_017009604.1 | Intron | XP_016865093.1 | ||||
XM_017009605.1 | Intron | XP_016865094.1 |