Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAACCCCCAACCCTGTCAATAA[A/G]CCACGAAAACTAGGAAGGCACGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609986 MIM: 604181 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD6 - caspase recruitment domain family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100506548 - uncharacterized LOC100506548 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL37 - ribosomal protein L37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000997.4 | 1197 | UTR 3 | NP_000988.1 |
SNORD72 - small nucleolar RNA, C/D box 72 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |