Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGTTTCTAGGAATTCCATATAA[A/G]CAACTGACTGTTGGAGTCCCCAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607878 | ||||||||||||||||||||
Literature Links: |
NNT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NNT - nicotinamide nucleotide transhydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012343.3 | 386 | Silent Mutation | AAA,AAG | K,K 55 | NP_036475.3 | |
NM_182977.2 | 386 | Silent Mutation | AAA,AAG | K,K 55 | NP_892022.2 | |
XM_005248274.4 | 386 | Silent Mutation | AAA,AAG | K,K 55 | XP_005248331.1 | |
XM_005248275.4 | 386 | Intron | XP_005248332.1 | |||
XM_006714461.3 | 386 | Intron | XP_006714524.1 | |||
XM_011514001.2 | 386 | Silent Mutation | AAA,AAG | K,K 55 | XP_011512303.1 | |
XM_011514002.2 | 386 | Intron | XP_011512304.1 | |||
XM_017009293.1 | 386 | Silent Mutation | AAA,AAG | K,K 55 | XP_016864782.1 |
NNT-AS1 - NNT antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |