Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGGCTATGGCTCCAGTGCCAC[A/G]TGCTTTGGCTCCAACCTGTCTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123295 MIM: 606614 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKMT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKMT2 - creatine kinase, mitochondrial 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099735.1 | Intron | NP_001093205.1 | ||||
NM_001099736.1 | Intron | NP_001093206.1 | ||||
NM_001825.2 | Intron | NP_001816.2 |
CKMT2-AS1 - CKMT2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRF2 - Ras protein specific guanine nucleotide releasing factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |