Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGATGAAAATTTGATGCTGAA[C/G]AAAGAACTCCATTTCCCCTGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616203 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC38A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC38A9 - solute carrier family 38 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258286.1 | Intron | NP_001245215.1 | ||||
NM_001258287.1 | Intron | NP_001245216.1 | ||||
NM_001282429.1 | Intron | NP_001269358.1 | ||||
NM_173514.3 | Intron | NP_775785.2 | ||||
XM_006714537.2 | Intron | XP_006714600.1 | ||||
XM_006714538.3 | Intron | XP_006714601.1 | ||||
XM_006714539.3 | Intron | XP_006714602.1 | ||||
XM_011543173.1 | Intron | XP_011541475.1 | ||||
XM_011543174.1 | Intron | XP_011541476.1 | ||||
XM_011543176.1 | Intron | XP_011541478.1 | ||||
XM_011543177.1 | Intron | XP_011541479.1 | ||||
XM_011543178.1 | Intron | XP_011541480.1 | ||||
XM_011543180.1 | Intron | XP_011541482.1 | ||||
XM_011543182.1 | Intron | XP_011541484.1 | ||||
XM_011543183.1 | Intron | XP_011541485.1 | ||||
XM_011543184.1 | Intron | XP_011541486.1 | ||||
XM_017009077.1 | Intron | XP_016864566.1 | ||||
XM_017009078.1 | Intron | XP_016864567.1 | ||||
XM_017009079.1 | Intron | XP_016864568.1 | ||||
XM_017009080.1 | Intron | XP_016864569.1 | ||||
XM_017009081.1 | Intron | XP_016864570.1 | ||||
XM_017009082.1 | Intron | XP_016864571.1 |