Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTTCGCTGTACACCCGGAGCC[C/T]GAGGCTTGCGGCGACCACGAGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607811 | ||||||||||||||||||||
Literature Links: |
C6orf52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf52 - chromosome 6 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145020.1 | 484 | Intron | NP_001138492.1 | |||
XM_006715088.3 | 484 | Intron | XP_006715151.1 | |||
XM_011514572.2 | 484 | Intron | XP_011512874.1 | |||
XM_011514574.2 | 484 | Intron | XP_011512876.1 | |||
XM_011514575.2 | 484 | Intron | XP_011512877.1 |
PAK1IP1 - PAK1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017906.2 | 484 | Silent Mutation | CCC,CCT | P,P 21 | NP_060376.2 | |
XM_005249204.2 | 484 | Silent Mutation | CCC,CCT | P,P 21 | XP_005249261.1 | |
XM_011514720.1 | 484 | Intron | XP_011513022.1 | |||
XM_011514721.1 | 484 | Intron | XP_011513023.1 |