Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGGGTAGAAGTATTTCACCTG[A/C]CCCATGAAGAACCTGATGGCTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEDD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEDD9 - neural precursor cell expressed, developmentally down-regulated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142393.1 | 3940 | UTR 3 | NP_001135865.1 | |||
NM_001271033.1 | 3940 | UTR 3 | NP_001257962.1 | |||
NM_006403.3 | 3940 | UTR 3 | NP_006394.1 | |||
NM_182966.3 | 3940 | Intron | NP_892011.2 |