Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTCTTCAAACGCCAGTCCAGAA[A/G]CATAGGGACAGCTTCACTACAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDN1 - endothelin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168319.1 | Intron | NP_001161791.1 | ||||
NM_001955.4 | Intron | NP_001946.3 | ||||
XM_011514330.2 | Intron | XP_011512632.1 | ||||
XM_011514331.2 | Intron | XP_011512633.1 | ||||
XM_011514332.2 | Intron | XP_011512634.1 | ||||
XM_017010331.1 | Intron | XP_016865820.1 |