Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCGTGCTTCTGCCTAAGAAGAC[C/G]GAGAGTCACCACAAGGCCAAGGGCA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 602794 MIM: 602847 | |||||||||||||||||||||||||||||
Literature Links: |
HIST1H2AC PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
AFR
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
SAS - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
HIST1H2AC - histone cluster 1, H2ac | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2BC - histone cluster 1, H2bc | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |