Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTGTCAGCTGGCTCTGTACTTC[C/T]ATTGGGCTGGTTCCTTCACCATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615932 MIM: 606964 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD20 - potassium channel tetramerization domain containing 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK38 - serine/threonine kinase 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305102.1 | 2726 | UTR 3 | NP_001292031.1 | |||
NM_007271.3 | 2726 | UTR 3 | NP_009202.1 | |||
XM_006714988.3 | 2726 | UTR 3 | XP_006715051.1 | |||
XM_006714989.3 | 2726 | UTR 3 | XP_006715052.1 | |||
XM_017010226.1 | 2726 | Intron | XP_016865715.1 |