Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAAGAACTATTTTTTGGCCTAA[C/T]GTGTATCAGGGACAGTGCATCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM33 - RNA binding motif protein 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053043.2 | Intron | NP_444271.2 | ||||
XM_005249531.1 | Intron | XP_005249588.1 | ||||
XM_005249532.1 | Intron | XP_005249589.1 | ||||
XM_005249533.1 | Intron | XP_005249590.1 | ||||
XM_005249534.1 | Intron | XP_005249591.1 | ||||
XM_005249535.1 | Intron | XP_005249592.1 | ||||
XM_011515840.1 | Intron | XP_011514142.1 |