Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGCTTTGGGTTTTGTTTTTTT[G/T]TTTTTTTTTTTTTTCAGTGGCATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604786 | ||||||||||||||||||||
Literature Links: |
ARL4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL4A - ADP ribosylation factor like GTPase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037164.2 | 2575 | UTR 3 | NP_001032241.1 | |||
NM_001195396.1 | 2575 | UTR 3 | NP_001182325.1 | |||
NM_005738.4 | 2575 | UTR 3 | NP_005729.1 | |||
NM_212460.3 | 2575 | UTR 3 | NP_997625.1 |