Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACCCACGTAGCTGAGGATGAC[A/G]GGCAGGAAGACCAAGCCATGCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPC1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPC1L1 - NPC1 like intracellular cholesterol transporter 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101648.1 | 3432 | Silent Mutation | NP_001095118.1 | |||
NM_001300967.1 | 3432 | Intron | NP_001287896.1 | |||
NM_013389.2 | 3432 | Silent Mutation | NP_037521.2 | |||
XM_011515326.2 | 3432 | Silent Mutation | XP_011513628.1 | |||
XM_011515327.2 | 3432 | Intron | XP_011513629.1 | |||
XM_011515328.2 | 3432 | Silent Mutation | XP_011513630.1 |