Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATAGTGCCTATGCAAGGTACCAG[C/G]CTCCTCAGAAGTCCTTGATGTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00525 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00525 - long intergenic non-protein coding RNA 525 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKD1L1 - polycystin 1 like 1, transient receptor potential channel interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138295.3 | 8589 | UTR 3 | NP_612152.1 | |||
XM_017011798.1 | 8589 | UTR 3 | XP_016867287.1 |