Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTGGGGCAGAACCTCAGATCA[C/T]AATCAGAAAACCACGCTCCTGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCF2 - ATP binding cassette subfamily F member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005692.4 | 3061 | Intron | NP_005683.2 | |||
NM_007189.2 | 3061 | UTR 3 | NP_009120.1 | |||
XM_017011639.1 | 3061 | Intron | XP_016867128.1 | |||
XM_017011640.1 | 3061 | Intron | XP_016867129.1 |
IQCA1L - IQ motif containing with AAA domain 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |