Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGGCTGGCTGCGAGGGGTACAC[A/G]GATCCCCGGTAGAAGGCGGAGCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608606 MIM: 614781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHLHA15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BHLHA15 - basic helix-loop-helix family member a15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TECPR1 - tectonin beta-propeller repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015395.2 | 3342 | Silent Mutation | TCC,TCT | S,S 1014 | NP_056210.1 | |
XM_005250253.3 | 3342 | Silent Mutation | TCC,TCT | S,S 1014 | XP_005250310.1 | |
XM_017011937.1 | 3342 | Silent Mutation | TCC,TCT | S,S 980 | XP_016867426.1 | |
XM_017011938.1 | 3342 | Intron | XP_016867427.1 |