Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCCGAGTTGAGCTGGACACAC[A/G]GAGCTCAGGGAGGCAGCAGTGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605341 MIM: 605342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PILRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PILRA - paired immunoglobin like type 2 receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013439.2 | 607 | Missense Mutation | CAG,CGG | Q,R 132 | NP_038467.2 | |
NM_178272.1 | 607 | Missense Mutation | CAG,CGG | Q,R 132 | NP_840056.1 | |
NM_178273.1 | 607 | Missense Mutation | CAG,CGG | Q,R 132 | NP_840057.1 |
PILRB - paired immunoglobin-like type 2 receptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAG3L5P-PVRIG2P-PILRB - STAG3L5P-PVRIG2P-PILRB readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |