Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGCCGGAGTTGAAGTCGGAGAG[C/G]GGGCAGACCGTCCAGGGTCAGGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611478 | ||||||||||||||||||||
Literature Links: |
MEPCE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEPCE - methylphosphate capping enzyme | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R35 - protein phosphatase 1 regulatory subunit 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145030.2 | 526 | Missense Mutation | CCT,GCT | P,A 228 | NP_659467.1 | |
XM_011515914.1 | 526 | Missense Mutation | CCT,GCT | P,A 158 | XP_011514216.1 |
ZCWPW1 - zinc finger CW-type and PWWP domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |