Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCAGCTGATGTTTGAGGGAACA[C/T]GGGGTAGCACTGCCTACCTGGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602372 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZAN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZAN - zonadhesin (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003386.2 | 709 | Missense Mutation | CGG,TGG | R,W 182 | NP_003377.2 | |
NM_173059.2 | 709 | Missense Mutation | CGG,TGG | R,W 182 | NP_775082.2 | |
XM_011516555.2 | 709 | Missense Mutation | CGG,TGG | R,W 182 | XP_011514857.1 | |
XM_017012581.1 | 709 | Intron | XP_016868070.1 |