Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAATTAGGCCTCCAAATCAAAAT[A/C]ATTTATATTAACCAAAAGGTTATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREB3L2 - cAMP responsive element binding protein 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253775.1 | 4747 | Intron | NP_001240704.1 | |||
NM_001318246.1 | 4747 | UTR 3 | NP_001305175.1 | |||
NM_194071.3 | 4747 | UTR 3 | NP_919047.2 | |||
XM_005250541.4 | 4747 | UTR 3 | XP_005250598.1 | |||
XM_017012540.1 | 4747 | UTR 3 | XP_016868029.1 | |||
XM_017012541.1 | 4747 | UTR 3 | XP_016868030.1 |