Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGATCATGGATACATGCAGTCA[A/G]TATGATGTATCCCTGTTGACGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613841 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBN2 - ubinuclein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173569.3 | Intron | NP_775840.3 | ||||
XM_005250249.3 | Intron | XP_005250306.2 | ||||
XM_006715916.3 | Intron | XP_006715979.2 | ||||
XM_011516002.2 | Intron | XP_011514304.1 | ||||
XM_011516003.1 | Intron | XP_011514305.1 |