Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACATTCCCTGGCAGCCTCGCAGC[C/T]TCGGGCAGATGGAGCTTCCGGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611432 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf66 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf66 - chromosome 9 open reading frame 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152569.2 | 1301 | Silent Mutation | GAA,GAG | E,E 268 | NP_689782.2 |
DOCK8 - dedicator of cytokinesis 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190458.1 | 1301 | Intron | NP_001177387.1 | |||
NM_001193536.1 | 1301 | Intron | NP_001180465.1 | |||
NM_203447.3 | 1301 | Intron | NP_982272.2 | |||
XM_011518045.2 | 1301 | Intron | XP_011516347.1 | |||
XM_011518046.2 | 1301 | Intron | XP_011516348.1 | |||
XM_011518047.2 | 1301 | Intron | XP_011516349.1 | |||
XM_011518048.2 | 1301 | Intron | XP_011516350.1 | |||
XM_011518049.2 | 1301 | Intron | XP_011516351.1 | |||
XM_017015173.1 | 1301 | Intron | XP_016870662.1 | |||
XM_017015174.1 | 1301 | Intron | XP_016870663.1 |