Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAACTAGGGAGAGAAAGTAAA[C/T]TCAACAGAAGGAGGGCCCAGGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600085 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYK - spleen associated tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135052.3 | Intron | NP_001128524.1 | ||||
NM_001174167.2 | Intron | NP_001167638.1 | ||||
NM_001174168.2 | Intron | NP_001167639.1 | ||||
NM_003177.6 | Intron | NP_003168.2 | ||||
XM_005252147.3 | Intron | XP_005252204.1 | ||||
XM_011518946.2 | Intron | XP_011517248.1 |