Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTTCTAGGGGGTGTGAATCTCC[C/T]GTTGGTCTTCTGTGACAGAAAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC613206 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC613206 - myeloproliferative disease associated tumor antigen 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK6 - NIMA related kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145001.2 | Intron | NP_001138473.1 | ||||
NM_001166167.1 | Intron | NP_001159639.1 | ||||
NM_001166168.1 | Intron | NP_001159640.1 | ||||
NM_001166169.1 | Intron | NP_001159641.1 | ||||
NM_001166170.1 | Intron | NP_001159642.1 | ||||
NM_001166171.1 | Intron | NP_001159643.1 | ||||
NM_014397.5 | Intron | NP_055212.2 | ||||
XM_005251664.1 | Intron | XP_005251721.1 | ||||
XM_006716936.2 | Intron | XP_006716999.1 | ||||
XM_017014217.1 | Intron | XP_016869706.1 |