Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCTGCTGCTGAGCGGCCGCCCC[A/G]CGCGCGCCGACAGTAAGGTACGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602778 MIM: 615899 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR6A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NR6A1 - nuclear receptor subfamily 6 group A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OLFML2A - olfactomedin like 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282715.1 | 186 | Intron | NP_001269644.1 | |||
NM_182487.3 | 186 | Missense Mutation | ACG,GCG | T,A 25 | NP_872293.2 | |
XM_005251760.4 | 186 | Missense Mutation | ACG,GCG | T,A 25 | XP_005251817.1 | |
XM_006716989.2 | 186 | Missense Mutation | ACG,GCG | T,A 25 | XP_006717052.1 |