Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCCCGCAGAAATGCCCCTCCGG[C/G]CTAGGAGCGACGGCGCCGGCCCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612898 MIM: 610727 | ||||||||||||||||||||
Literature Links: |
COQ4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COQ4 - coenzyme Q4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305942.1 | 436 | Missense Mutation | CCT,GCT | P,A 29 | NP_001292871.1 | |
NM_016035.4 | 436 | Missense Mutation | CCT,GCT | P,A 29 | NP_057119.2 | |
XM_017014792.1 | 436 | Missense Mutation | CCT,GCT | P,A 29 | XP_016870281.1 | |
XM_017014793.1 | 436 | Missense Mutation | CCT,GCT | P,A 29 | XP_016870282.1 |
TRUB2 - TruB pseudouridine synthase family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |