Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCTGCTTGGAAGGTCAGCTCT[C/G]AGAGAGCTGGTAAGTGGTGGTGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610746 MIM: 615587 | ||||||||||||||||||||
Literature Links: |
DOLK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOLK - dolichol kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP188 - nucleoporin 188 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015354.2 | 105 | Silent Mutation | CTC,CTG | L,L 26 | NP_056169.1 |
PHYHD1 - phytanoyl-CoA dioxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |