Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTTAGCCTGGTTAAGATGCC[A/G]TTTTCTTTCTCCTTCGCCTCCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600789 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL23 - mitochondrial ribosomal protein L23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021134.3 | Intron | NP_066957.3 | ||||
XM_006718271.2 | Intron | XP_006718334.1 | ||||
XM_011520273.1 | Intron | XP_011518575.1 | ||||
XM_011520275.2 | Intron | XP_011518577.1 | ||||
XM_011520276.2 | Intron | XP_011518578.1 |
SNORD131 - small nucleolar RNA, C/D box 131 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |