Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCAGTCCAGATCGACCAGGCG[C/T]GGCAGAGAGACTGTGGAAGGAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612299 | ||||||||||||||||||||
Literature Links: |
COMMD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COMMD9 - COMM domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101653.1 | 365 | Silent Mutation | CCA,CCG | P,P 79 | NP_001095123.1 | |
NM_001307932.1 | 365 | Missense Mutation | ACG,GCG | T,A 110 | NP_001294861.1 | |
NM_001307937.1 | 365 | Silent Mutation | CCA,CCG | P,P 112 | NP_001294866.1 | |
NM_014186.3 | 365 | Silent Mutation | CCA,CCG | P,P 121 | NP_054905.2 | |
XM_017017625.1 | 365 | Intron | XP_016873114.1 |