Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTCAGTGCTTCCAAGATTAGAT[C/T]GGGAGTTTTCTTCATGAGGTCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
API5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
API5 - apoptosis inhibitor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142930.1 | Intron | NP_001136402.1 | ||||
NM_001142931.1 | Intron | NP_001136403.1 | ||||
NM_001243747.1 | Intron | NP_001230676.1 | ||||
NM_006595.3 | Intron | NP_006586.1 | ||||
XM_006718359.3 | Intron | XP_006718422.1 | ||||
XM_017018464.1 | Intron | XP_016873953.1 | ||||
XM_017018465.1 | Intron | XP_016873954.1 |