Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACATTCCACAGGATAAGGAAG[A/G]AAAACTGTGAAATCACTTTGAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
AEBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AEBP2 - AE binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114176.1 | Intron | NP_001107648.1 | ||||
NM_001267043.1 | Intron | NP_001253972.1 | ||||
NM_153207.4 | Intron | NP_694939.2 | ||||
XM_006719031.3 | Intron | XP_006719094.1 | ||||
XM_011520560.2 | Intron | XP_011518862.1 | ||||
XM_017018806.1 | Intron | XP_016874295.1 |