Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATAAGTGAGTGACCCAAGGGGT[A/G]GAGAATTGCTGCAATCTCTCCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604793 MIM: 604794 MIM: 604795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAS2R7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAS2R7 - taste 2 receptor member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAS2R8 - taste 2 receptor member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023918.1 | 829 | Missense Mutation | CAC,TAC | H,Y 277 | NP_076407.1 |
TAS2R9 - taste 2 receptor member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |