Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAAACAAAGTGCTGGCTGGGAA[G/T]AATTCTTGAATGTGCATATCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600579 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRO - protein tyrosine phosphatase, receptor type O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002848.3 | Intron | NP_002839.1 | ||||
NM_030667.2 | Intron | NP_109592.1 | ||||
NM_030668.2 | Intron | NP_109593.1 | ||||
NM_030669.2 | Intron | NP_109594.1 | ||||
NM_030670.2 | Intron | NP_109595.1 | ||||
NM_030671.2 | Intron | NP_109596.1 | ||||
XM_017019725.1 | Intron | XP_016875214.1 |