Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGCCAAGCCCGCAGGAGACCC[A/C]ACGGCCCACCAGCCTTTCTGGGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605864 MIM: 605219 | ||||||||||||||||||||
Literature Links: |
B3GNT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B3GNT4 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011538732.1 | 232 | Silent Mutation | CCA,CCC | P,P 34 | XP_011537034.1 |
DIABLO - diablo IAP-binding mitochondrial protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC43 - leucine rich repeat containing 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |