Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTTCTTGCCTCTTACAAGGCA[C/G]CGCGTGTAACGTCCTGACCAGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX55 - DEAD-box helicase 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020936.2 | 1563 | Intron | NP_065987.1 | |||
XM_011538608.2 | 1563 | Intron | XP_011536910.1 | |||
XM_017019710.1 | 1563 | Intron | XP_016875199.1 | |||
XM_017019711.1 | 1563 | Intron | XP_016875200.1 | |||
XM_017019712.1 | 1563 | Intron | XP_016875201.1 | |||
XM_017019713.1 | 1563 | Intron | XP_016875202.1 | |||
XM_017019714.1 | 1563 | Intron | XP_016875203.1 | |||
XM_017019715.1 | 1563 | Intron | XP_016875204.1 | |||
XM_017019716.1 | 1563 | Intron | XP_016875205.1 | |||
XM_017019717.1 | 1563 | Intron | XP_016875206.1 | |||
XM_017019718.1 | 1563 | Intron | XP_016875207.1 | |||
XM_017019719.1 | 1563 | Intron | XP_016875208.1 | |||
XM_017019720.1 | 1563 | Intron | XP_016875209.1 | |||
XM_017019721.1 | 1563 | Intron | XP_016875210.1 |
EIF2B1 - eukaryotic translation initiation factor 2B subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001414.3 | 1563 | UTR 3 | NP_001405.1 |