Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTTCAGGGCGCCCTGCGTCCG[A/G]CAGAGGAGGCGAGCATCCCGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC92 - coiled-coil domain containing 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304957.1 | 154 | Intron | NP_001291886.1 | |||
NM_001304958.1 | 154 | Intron | NP_001291887.1 | |||
NM_001304959.1 | 154 | Intron | NP_001291888.1 | |||
NM_001304960.1 | 154 | Intron | NP_001291889.1 | |||
NM_001304961.1 | 154 | Intron | NP_001291890.1 | |||
NM_025140.2 | 154 | Intron | NP_079416.1 | |||
XM_005253624.2 | 154 | Intron | XP_005253681.1 | |||
XM_017019984.1 | 154 | Intron | XP_016875473.1 |
ZNF664 - zinc finger protein 664 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204298.1 | 154 | UTR 5 | NP_001191227.1 | |||
NM_152437.2 | 154 | UTR 5 | NP_689650.1 |
ZNF664-FAM101A - filamin-interacting protein FAM101A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204299.1 | 154 | UTR 5 | NP_001191228.1 |