Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGTGGAGTTGTTTGAGTTTAA[A/G]GGAGAAGATCTGACGGAAGAGGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600611 | ||||||||||||||||||||
Literature Links: |
FKBP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FKBP4 - FK506 binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002014.3 | 612 | Silent Mutation | AAA,AAG | K,K 138 | NP_002005.1 | |
XM_011520929.2 | 612 | Silent Mutation | AAA,AAG | K,K 93 | XP_011519231.1 |