Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATAGATGTGAGTGTGGGAGAAGA[C/T]CCCATATTTGGCTAAGGAGCACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf43 - chromosome 12 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OASL - 2'-5'-oligoadenylate synthetase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261825.1 | 1523 | Missense Mutation | ATC,GTC | I,V 288 | NP_001248754.1 | |
NM_003733.3 | 1523 | Missense Mutation | ATC,GTC | I,V 418 | NP_003724.1 | |
NM_198213.2 | 1523 | UTR 3 | NP_937856.1 | |||
XM_017020140.1 | 1523 | Intron | XP_016875629.1 | |||
XM_017020141.1 | 1523 | Intron | XP_016875630.1 |