Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGATGTGTTTTCAAGTTACCATC[C/T]TTAACTTCTAAGAGGTCACCTTTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612755 | ||||||||||||||||||||
Literature Links: |
MIR3657 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR3657 - microRNA 3657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA25 - N(alpha)-acetyltransferase 25, NatB auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024953.3 | 3199 | Silent Mutation | AAA,AAG | K,K 827 | NP_079229.2 | |
XM_005253938.1 | 3199 | Silent Mutation | AAA,AAG | K,K 691 | XP_005253995.1 | |
XM_006719606.2 | 3199 | Silent Mutation | AAA,AAG | K,K 799 | XP_006719669.1 | |
XM_017019977.1 | 3199 | Silent Mutation | AAA,AAG | K,K 476 | XP_016875466.1 |