Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGAACATGCCGTGGTACATGG[C/T]AAGGTACAAAGCCGTTAGAGCCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607568 MIM: 251170 | ||||||||||||||||||||
Literature Links: |
MMAB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMAB - methylmalonic aciduria (cobalamin deficiency) cblB type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVK - mevalonate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000431.3 | 221 | Silent Mutation | GGC,GGT | G,G 25 | NP_000422.1 | |
NM_001114185.2 | 221 | Silent Mutation | GGC,GGT | G,G 25 | NP_001107657.1 | |
NM_001301182.1 | 221 | Silent Mutation | GGC,GGT | G,G 25 | NP_001288111.1 | |
XM_011538372.2 | 221 | Silent Mutation | GGC,GGT | G,G 125 | XP_011536674.2 | |
XM_017019313.1 | 221 | Silent Mutation | GGC,GGT | G,G 25 | XP_016874802.1 | |
XM_017019314.1 | 221 | Silent Mutation | GGC,GGT | G,G 25 | XP_016874803.1 |