Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGACTGAGCCGGGCATGTTCTTC[A/G]GGCCCCTGTGTGAGTGCCATGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGBL1 - integrin subunit beta like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271754.1 | 455 | Intron | NP_001258683.1 | |||
NM_001271755.1 | 455 | Missense Mutation | AGG,GGG | R,G 86 | NP_001258684.1 | |
NM_001271756.1 | 455 | Intron | NP_001258685.1 | |||
NM_004791.2 | 455 | Missense Mutation | AGG,GGG | R,G 86 | NP_004782.1 | |
XM_005254100.4 | 455 | Missense Mutation | AGG,GGG | R,G 86 | XP_005254157.1 |