Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTCTCTGCTGTAGGTCACCC[A/G]CCATGTCGACCTTGCTGGACATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAIAP3 - BAI1 associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199096.1 | 756 | UTR 5 | NP_001186025.1 | |||
NM_001199097.1 | 756 | UTR 5 | NP_001186026.1 | |||
NM_001199098.1 | 756 | UTR 5 | NP_001186027.1 | |||
NM_001199099.1 | 756 | UTR 5 | NP_001186028.1 | |||
NM_001286464.1 | 756 | UTR 5 | NP_001273393.1 | |||
NM_003933.4 | 756 | Missense Mutation | ACC,GCC | T,A 35 | NP_003924.2 | |
XM_011522728.1 | 756 | Missense Mutation | ACC,GCC | T,A 35 | XP_011521030.1 | |
XM_011522729.1 | 756 | Missense Mutation | ACC,GCC | T,A 35 | XP_011521031.1 | |
XM_011522730.2 | 756 | Missense Mutation | ACC,GCC | T,A 35 | XP_011521032.1 |